missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Control Slides, IHC Total RB 3C8

All control slides are on Mopec™ IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.

$1422.37 - $3248.37

Specifications

Coating Uncoated
Thickness (Metric) 1 mm
Material Glass
Species Human
Tissue Condition Normal
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
22-448-35
View Documents
Mopec™
SC56025
24 Unstained, 1 Stained
Case of 25 Each for $1,422.37
Only null left
 
22-448-34
View Documents
Mopec™
SC560100
98 Unstained, 2 Stained
Case of 100 Each for $3,248.37
Only null left
 
Description

Description

  • Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
  • Case of 25 contains 24 unstained slides and 1 stained slide
  • Case of 100 contains 98 unstained slides and 2 stained slides
Specifications

Specifications

Uncoated
Glass
Normal
75 x 25 mm
1 mm
Human
IHC Control Slide
No
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'