missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Control Slides, IHC LAMA5

All control slides are on Mopec™ IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.

$3676.07 - $3676.07

Specifications

Coating Uncoated
Thickness (Metric) 1 mm
Material Glass
Species Human
Tissue Condition Normal
View More Specs

Products 1
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
22-445-12
View Documents
Mopec™
SC297100
98 Unstained, 2 Stained
Case of 100 Each for $3,676.07
Only null left
 
Description

Description

  • Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
  • Case of 25 contains 24 unstained slides and 1 stained slide
  • Case of 100 contains 98 unstained slides and 2 stained slides
Specifications

Specifications

Uncoated
Glass
Normal
75 x 25 mm
1 mm
Human
IHC Control Slide
No
Videos
SDS
Documents

Documents