missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Control Slides, IHC CD 117

Catalog No. 2245192
Change view
Click to view available options
Type:
Control Slides, IHC CD 117
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
2245192 Control Slides, IHC CD 117
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2245192 Supplier Mopec™ Supplier No. SC60025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

All control slides are on Mopec™ IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.

  • Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
  • Case of 25 contains 24 unstained slides and 1 stained slide
  • Case of 100 contains 98 unstained slides and 2 stained slides

Specifications

Coating Uncoated
Thickness (Metric) 1 mm
Includes 24 Unstained Slides, 1 Stained Slide
Material Glass
Species Human
Tissue Condition Normal
Type Control Slides, IHC CD 117
Dimensions (LxW) 75 x 25 mm
Enhanced Surface No