missing translation for 'onlineSavingsMsg'
Learn More

Mopec Control Slides, IHC, Box of 100, 98 Unstained, +2 Stained, Gastrin, Middle Placement

Catalog No. 22010175
Change view
Click to view available options
:
Control Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22010175 Control Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22010175 Supplier Mopec Supplier No. SC606100CUST
Only null left

Designed to support the precise demands of laboratories, Mopec Control Slides include Special Stain, IHC, Normal Human Tissue, and Tissue Microarray (TMA).

  • Mopec recognizes the crucial role control slides play in ensuring diagnostic accuracy and reliability, which is why we also provide a unique option for ordering custom control slides, ensuring a precise fit to your specifications.
  • Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80°F in a humidity controlled and light-deprived environment.
  • Please revalidate after one year to verify continued reactivity.
  • All control slides are on Mopec IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.

Specifications

Coating Uncoated
Thickness (Metric) 4 μm
Material Microscopic Glass
Species Human
Tissue Condition Normal
Type IHC Control Slide
Dimensions (LxW) 11 x 7 in.
Enhanced Surface No