missing translation for 'onlineSavingsMsg'
Learn More

Contec™ VertiKlean™ Sox Mop Head Covers
GREENER_CHOICE

Catalog No. 19159256
Change view
Click to view available options
Size:
Medium
For Use With (Equipment):
Medium VertiKlean Mop Head
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size For Use With (Equipment)
19159256 Medium Medium VertiKlean Mop Head
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19159256 Supplier Contec™ Supplier No. MVKQTSOX

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Increases durability of VertiKlean mop when used on abrasive surfaces. Contec™ VertiKlean™ Sox Mop Head Covers are available in regular, medium and large to fit all VertiKlean mop heads.

  • Clean-processed polyester
  • Extends the life of VertiKlean mop heads

Specifications

Color Code White
Product Type Mop Cover
Sterility Non-sterile
Autoclavable Not Autoclavable
Size Medium
Disposable Single-use
For Use With (Equipment) Medium VertiKlean Mop Head
For Use With (Application) Removable cover increases durability of VertiKlean mop when used on abrasive surfaces
Color-coded No
Material Polyester