Learn More
Contec™ TuffStuff™ Critical Task Wipes
Description
- Ideal for a variety of tough jobs including general industrial cleaning, production line and heavy equipment maintenance, and hand and tool cleanup
- Constructed of cellulose and polyester
- Can be used with harsh solvents such as isopropyl alcohol, MEK, and MPK without falling apart
- Contain no binders, chemical residue, or contaminants that can result in surface damage or rework
- Recommended for critical cleaning in biotech and pharmaceutical industries
Specifications
Specifications
| Product Type | Critical Task Wipe |
| Material | Polyester Cellulose |
| Certifications/Compliance | ISO Class 6-8 environments |
| Dimensions (L x W) | 12 x 13 in. |
| Format | Roll in Dispenser Box |
| Packaging Quantity | 500/Roll, 1 Roll/Dispenser |
| Sterility | Non-sterile |