missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Irradiated Polyethylene Bucket Liners

Catalog No. 19200000
Change view
Click to view available options
Thickness (Metric):
4 mil
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thickness (Metric)
19200000 4 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19200000 Supplier Contec™ Supplier No. BKTBAG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fits 5 gal. (20 L) and 6.5 gal. (25 L) and 7 gal. (27 L) buckets. Contec™ Bucket Liners protect buckets and disinfectant solutions by reducing the opportunity for cross contamination.

  • Intended for single use
  • Keeps solutions sterile, even without autoclave access
  • Manufactured and packaged in an ISO Class 4 cleanroom
  • Gamma irradiated with a specified dose of 25-50 kGy

Specifications

Product Type Bucket Liner
Material LDPE
Autoclavable Autoclavable
Color Clear
Disposable Single-use
For Use With (Equipment) 5 gal. (20 L), 6.5 gal. (25 L), 7 gal. (27 L) Buckets
Sterility Non-sterile
Thickness (Metric) 4 mil