Learn More
Contec™ Drip Caps for Multibucket Systems
Catalog No. 19168753
Change view
Click to view available options
For Use With (Equipment):
5 gal. multi-Bucket System
6.5 gal. multi-Bucket System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19168753 6.5 gal. multi-Bucket System
19159235 5 gal. multi-Bucket System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19168753 Supplier Contec™ Supplier No. 2729
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
A stainless-steel connecting kit to prevent drips between the buckets.
Fits Contec™ Bucket Systems. The Contec™ Stainless Steel Drip Cap is easy to install and prevents drips between buckets.
  • Compatible with all disinfectant solutions and common solvents
  • 100% stainless steel for durability and cleanliness
  • Autoclavable
Specifications
Autoclavable Autoclavable
For Use With (Equipment) 6.5 gal. multi-Bucket System