Learn More
Contec™ SATWIPES™ Presaturated Wipes
GREENER_CHOICE
Prepare for a variety of cleanrooms. The SATWIPES™Presaturated Wipes are offered in various options to help tackle general-purpose cleaning in ISO Class 5 to ISO Class 8 environments.
Supplier:  Contec™ SATC3694
 View more versions of this product
Catalog No. 19033401
Only null left
Description
Description
  • Durable, presaturated wipes with a choice of solvent saturations on perforated rolls
  • Polyester Nonwoven are ideal for critical applications in ISO Class 5 environments
  • SATWIPES with Polyester/Cellulose Nonwoven Wipes offer strength and low extractables for use in less critical applications in ISO Class 6 to 8 environments
  • Designed for general-purpose cleaning and surface preparation and for wiping process tools and other equipment and for use prior to cleanroom entry
Order Info
Packaged one hundred 6 x 9 in. (15 x 23cm) wipers on a perforated roll. Most SATWipes cases contain 12 presaturated rolls plus one canister; 19-150-0711 and -0712 packed 18 refills per case without a canister. Custom-blend substrates and additional wiper sizes available through your Fisher Safety Customer Service Representative. Empty canisters for wipes is available: 19-171-955.
Specifications
Chemical Identifiers
Pre-saturated Wipe
Not Autoclavable
Single-use
Non-sterile
Specifications
Pre-saturated Wipe
22.9 cm
15.2 cm
Not Autoclavable
Clear
23 x 15 cm (9 x 6 in.)
12 Pk.
6% IPA, 94% DI Water
9 in.
6 in.
Polyester
ISO Class 6
Single-use
Excellent wipes for less critical applications, compatible with ISO class 6 environments and wipes down prior to entry into cleanroom
1-ply
Non-sterile
Product Suggestions
Product Suggestions