missing translation for 'onlineSavingsMsg'
Learn More

Contec™ EasySat™ Bucketless Floor Mop Heads
GREENER_CHOICE

Catalog No. 191500694
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
191500694 Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 191500694 Supplier Contec™ Supplier No. BLVKMOPIR

Eliminate the need for a bucket or wringer in cleanroom mopping. The Contec™ EasySat™ Bucketless Floor Mop features laser-cut sealed-edge VertiKlean™ mop head material and is fully autoclavable. A push button flow control releases cleaning solution on demand.

Contec's EasySat™ bucketless floor mop is an effective and quick solution to your cleaning challenges. Designed for the cleanroom, this innovative mop is easy to use –– the built-in 52 ounce solution tank completely eliminates the need for a bucket and wringer. Single-use mop heads are made from our ultra clean polyester/ester foam VertiKlean™ material and attach to hardware in seconds using traditional hook and loop fasteners.

The angled mop head cleans hard-to-reach areas and is available in two styles: smooth surface for standard floor cleaning, and textured surface for more abrasive action. The mop head is available Validated Sterile - gamma irradiated to 10-6 SAL.

Features & Benefits

  • Hydrophilic polyester/clean ester, laser cut sealed edge foam material
  • Translucent tank allows user to see solution
  • Fully autoclavable for critical areas
  • Easy to use
  • Compatible with a wide range of solvents

Specifications

Dimensions 21.5 x 4.75 x 18.25 in.
Product Type Mop Head
Sterility Sterile
Autoclavable Autoclavable
Disposable Single-use
For Use With (Equipment) BLMOP
Color-coded No
Material Knitted Polyester, Ester Foam, Polyester Loop Fabric