missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Connector Cables for Automated Thermal Cycler (ATC)

Catalog No. A31482
Encompass
Change view
Click to view available options
Format:
1-m connector cable (flexible)
3-m connector cable (flexible)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
A31482 1-m connector cable (flexible)
A31483 3-m connector cable (flexible)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31482 Supplier Applied Biosystems™ Supplier No. A31482

Includes

1 meter connection cable for Automated Thermal Cycler (ATC)

Used to connect the Automated Thermal Cycler (ATC) block to the power box, this connector is available in a rigid 10-cm, flexible 1-m, or flexible 3-m cable format.

For customers seeking to automate their PCR workflow, the Applied Biosystems Automated Thermal Cycler (ATC) provides the reliability and performance you've come to expect from Applied Biosystems instruments. Small, easy-to-integrate, with a compatible format, our Automated Thermal Cycler enables hands-free PCR results.

In order to better align with lab design and demands, a longer or shorter cable may be utilized to optimize workflow.

Warranty

27 month warranty

Specifications

Format 1-m connector cable (flexible)
For Use With (Equipment) Automated Thermal Cycler (ATC)
Product Type Connector Cable
For Use With (Application) PCR
Max. Heating Rate (Thermal Block) 2.8°C/sec. (Block), 1.6°C/sec. (Sample)
Quantity 1 cable

For Research Use Only. Not for use in diagnostic procedures.