Learn More
Connecticut Clean Room™ PolyTack Roll Mop and Accessories
Perfect for cleaning walls, floors, ceiling and hard to reach areas
Supplier: Connecticut Clean Room™ PR9F
Description
- Helps keep areas free of contamination with easy to use hand or pole attachments
- Special anti-microbial agents provide long-lasting protection against bacterial growth
- Plastic inner core insures cleanliness
- Refills and replacement handles, mops and tacky rollers available
- 19-823-738 is a film roll only; it does not include a handle or extension pole
Chemical Identifiers
| For Removing Particulate Matter from the Cleanroom |
Specifications
| Tacky Roller | |
| Roll Mop | |
| Plastic Inner Core Foam |
| 22.8 cm (9 in.) | |
| For Removing Particulate Matter from the Cleanroom |