missing translation for 'onlineSavingsMsg'
Learn More

Connecticut Clean Room™ PolyTack Roll Mop and Accessories

Catalog No. 19823737
Encompass
Change view
Click to view available options
Material:
Metal
Plastic Inner Core Foam
Product Type:
Roller Mop Handle
Tacky Roller
Size:
22.8 cm (9 in.)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type Size
19823737 Metal Roller Mop Handle 22.8 cm (9 in.)
19823739 Plastic Inner Core Foam Tacky Roller 22.8 cm (9 in.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19823737 Supplier Connecticut Clean Room™ Supplier No. PH9
Only null left

Perfect for cleaning walls, floors, ceiling and hard to reach areas

 

  • Helps keep areas free of contamination with easy to use hand or pole attachments
  • Special anti-microbial agents provide long-lasting protection against bacterial growth
  • Plastic inner core insures cleanliness
  • Refills and replacement handles, mops and tacky rollers available
  • 19-823-738 is a film roll only; it does not include  a handle or extension pole

 

Specifications

Product Type Roller Mop Handle
Length (English) 18 in.
Size 22.8 cm (9 in.)
For Use With (Equipment) Tacky Roller Refill
For Use With (Application) For Removing Particulate Matter from the Cleanroom
Material Metal