Learn More
Thermo Scientific™ Connect Edge Independent Sensors for InstrumentConnect Remote Monitoring and OPC UATM
Thermo Scientific Connect Edge independent sensors and sensor kits are designed to collect data from laboratory equipment. Independent sensors can also be used as a redundant source of data in alongside Device Adapters. Sensors come with Standard or Accredited calibration.
$185.50 - $3,346.00
| Catalog Number | Mfr. No. | Calibration | IP Rating | Includes | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Calibration | IP Rating | Includes | Price | Quantity & Availability | |||
|
11731068
|
Thermo Scientific™
EDGEPTSENDU004 |
Not Calibrated | None | Dry Contact Door Switch Sensor with 3.7m cable for -25°C to 70°C (-15°F to 160°F) conditions |
Each for $185.50
|
|
|||
|
11731067
|
Thermo Scientific™
EDGEPTSENAU005 |
Not Calibrated | IP66/68 | Current Transducer 50A with 3.7m cable for -10°C to 70°C (14°F to 158°F) conditions |
Each for $210.33
|
|
|||
|
11731065
|
Thermo Scientific™
EDGEKTSENRA002 |
Standardized processed according to NIST and ISO/CEI 17025 | IP65 | Temperature/RH Sensor: -40° C to +60°C, Accredited Calibration: 37°C at 90% RH, includes 24V round cable (3.7m) |
Each for $1,481.80
|
|
|||
|
11731042
|
Thermo Scientific™
EDGEKTSENRC002 |
Standardized processed according to NIST and ISO9001 | IP65 | Temperature/RH Sensor: -40°C to +60°C, Standard Calibration: 37° C at 90% RH, includes 24V round cable (3.7m) |
Each for $1,386.20
|
|
|||
|
11731064
|
Thermo Scientific™
EDGEKTSENCA003 |
Standardized processed according to NIST and ISO/CEI 17025 | IP66/68 | CO2 Sensor with 24 V round cable (3.7 m), Accredited 23°C (with compensation for 37°C). See TDS for mor information. |
Each for $1,918.83
|
|
|||
|
11731066
|
Thermo Scientific™
EDGEPTSENTC006A |
Not Calibrated | IP66/68 | Digital Temperature Sensor: -55 to 125°C, CAL B, Standard Calibration: -40°C, -20°C, 0°C, includes 3.7 m flat PFTE cable. Standardized processed according to NIST and ISO9001. |
Each for $396.07
|
|
|||
|
11731059
|
Thermo Scientific™
EDGEPTSENTA006A |
Standardized processed according to NIST and ISO/CEI 17025 | IP66/68 | Digital Temperature Sensor: -55 to 125°C, CAL B, Accredited Calibration: -40°C, -20°C, 0°C, includes 3.7 m flat PFTE cable. Standardized processed according to NIST and ISO9001. |
Each for $518.98
|
|
|||
|
11731043
|
Thermo Scientific™
EDGEPTSENTC006B |
Standardized processed according to NIST and ISO9001 | IP68 | Digital Temperature Sensor: -55 to 125°C, CAL B, Standard Calibration: 0°C, 37°C, 100° C, includes 3.7 m flat PFTE cable. |
Each for $396.07
|
|
|||
|
11731061
|
Thermo Scientific™
EDGEPTSENTA006B |
Standardized processed according to NIST and ISO/CEI 17025 | IP66/68 | Digital Temperature Sensor: -55 to 125°C, CAL B, Accredited Calibration: 0°C, 37°C, 100°C, includes 3.7m flat PFTE cable. |
Each for $518.98
|
|
|||
|
11731044
|
Thermo Scientific™
EDGEPTSENRA007A |
Standardized processed according to NIST and ISO/CEI 17025 | IP66/68 | Digital Temperature/Relative Humidity Sensor: -40 to 125°C Standard Calibration: 23°C at 50% RH, includes 3 m round cable. |
Each for $434.76
|
|
|||
|
11731027
|
Thermo Scientific™
EDGEPTSENRC007A |
Standardized processed according to NIST and ISO9001 | IP66/68 | Digital Temperature/Relative Humidity Sensor: -40 to 125°C Standard Calibration: 23°C at 50% RH, includes 3 m round cable. |
Each for $336.87
|
|
|||
|
11731026
|
Thermo Scientific™
EDGEPTSENRA007B |
Standardized processed according to NIST and ISO/CEI 17025 | IP66/68 | Digital Temperature/Relative Humidity Sensor: -40 to 125°C Standard Calibration: 37°C at 90% RH, includes 3 m round cable |
Each for $434.76
|
|
|||
|
11731028
|
Thermo Scientific™
EDGEPTSENRC007B |
Standardized processed according to NIST and ISO9001 | IP66/68 | Digital Temperature/Relative Humidity Sensor: -40 to 125°C Standard Calibration: 23°C at 50% RH, includes 3 m round cable |
Each for $336.87
|
|
|||
|
11731038
|
Thermo Scientific™
EDGEPTSENTC001 |
Standardized processed according to NIST and ISO9001 | IP66/68 | PT100 4-20mA Sensor: -200 to 200°C, Class B, Standard Calibration: -196°C, -80°C, 0°C, includes 3.7 m round cable |
Each for $1,208.65
|
|
|||
|
11731058
|
Thermo Scientific™
EDGEPTSENTA001 |
Standardized processed according to NIST and ISO/CEI 17025 | IP66/68 | PT100 4-20mA Sensor: -200 to 200°C, Class B, Accredited Calibration: -196°C, -80°C, 0°C, includes 3.7 m round cable |
Each for $1,333.85
|
|
|||
|
11731040
|
Thermo Scientific™
EDGEPTSENTC008 |
Standardized processed according to NIST and ISO9001 | IP66/68 | PT100 4-20mA Sensor: -100 to 450°C, Class A, Standard Calibration -80°C, -40°C, 0°C, includes 3.7 m round cable |
Each for $1,333.85
|
|
|||
|
11731025
|
Thermo Scientific™
EDGEPTSENTA008 |
Standardized processed according to NIST and ISO/CEI 17025 | IP66/68 | PT100 4-20mA Sensor: -100 to 450°C, Class A, Accredited Calibration -80°C, -40°C, 0°C, includes 3.7 m round cable |
Each for $1,456.76
|
|
|||
|
11731048
|
Thermo Scientific™
EDGEKTARHC |
Standardized processed according to NIST and ISO/CEI 17025 | IP66/68 | T/RH/CO2 Kit: Includes Temperature /Relative Humidity Sensor with 24 V round cable (3 m), CO2 Sensor with round cable (3.7 m), Accredited Calibration: 37°C at 90% RH |
Each for $3,346.00
|
|
|||
|
11731049
|
Thermo Scientific™
EDGEKTCRHC |
Standardized processed according to NIST and ISO9001 | IP66/68 | T/RH/CO2 Kit: Includes Temperature /Relative Humidity Sensor with 24 V round cable (3 m), CO2 Sensor with round cable (3.7 m), Standard Calibration: 37°C at 90% RH |
Each for $3,323.24
|
|
|||
|
11731073
|
Thermo Scientific™
EDGEKTCCLD |
Standardized processed according to NIST and ISO9001 | IP66/68 | Cold Kit: Includes 4-20mA PT100; -100°C to +316°C (3.7 m round cable), dry contact door switch, 50A Current Transducer, Standard Calibration: -80°C, -40°C, 0°C |
Each for $1,727.63
|
|
|||
|
11731072
|
Thermo Scientific™
EDGEKTACLD |
Standardized processed according to NIST and ISO/CEI 17025 | IP66/68 | Cold Kit: Includes 4-20mA PT100; -100°C to +316°C (3.7 m round cable), dry contact door switch, 50A Current Transducer, Accredited Calibration: -80°C, -40°C, 0°C |
Each for $1,850.54
|
|
|||
|
11731033
|
Thermo Scientific™
EDGEKTALN2 |
Standardized processed according to NIST and ISO/CEI 17025 | IP66/68 | LN2 Kit: Includes 4-20mA PT100; 200°C to +200°C (3.7 m round cable) and dry contact door switch, Accredited Calibration: -196°C, -80°C, 0°C |
Each for $1,518.22
|
|
|||
|
11731077
|
Thermo Scientific™
EDGEKTCLN2 |
Standardized processed according to NIST and ISO9001 | IP66/68 | LN2 Kit: Includes 4-20mA PT100; 200°C to +200°C (3.7 m round cable) and dry contact door switch, Standard Calibration: -196°C, -80°C, 0°C |
Each for $1,393.02
|
|
|||
Description
Thermo Scientific™ InstrumentConnect is professional-grade remote monitoring system for monitoring laboratory equipment and other devices to help protect samples and increase asset management efficiencies. The InstrumentConnect system utilizes Connect Edge adapters and gateways to send data to the InstrumentConnect cloud or to your own premise-based system (LIMS, BMS, SCADA) using OPC-UA™ protocol. Connect Edge hardware is compatible with 700+ Thermo Fisher Scientific laboratory equipment makes and models.
System Hardware
- Device Adapter-Palm-size adapters collect and transmit data from lab equipment/devices to gateways using Ethernet (WAN) connectivity or Wi-Fi (where permitted).
- Sensor Adapter-Adapter to collect and transmit data from lab any OEM equipment/device to gateways using Ethernet (WAN) connectivity or Wi-Fi (where permitted).
- Ethernet Base Converter-used to convert any Wi-Fi device adapter to Ethernet/PoE. Used to convert Sensor Adapter to Ethernet (excludes PoE).
- Connect Edge Gateway-Receives data from adapters to send to InstrumentConnect (cloud) or customer on-premise system/cloud via OPC-UA using Ethernet (WAN) connectivity or Wi-Fi (where permitted).
System Software
Choose from these available InstrumentConnect packages.
InstrumentConnect Monitor (Cloud):
- Monitor equipment via advanced onboard equipment sensors
- Receive alarms and notifications, set custom alarms
- View live device status 24/7 via Mobile and Cloud app
- Peace of mind with industry-leading security protocols
- 25GB included data storage for documents
- Unlimited device telemetry storage
- Notifications via mobile application, e-mail and desktop application
instrumentConnect Integrate for OPC UA (OPC-UA to Cloud or Prem)
- Utilize the Connect Edge system to collect and transmit data to your on-premises system or cloud using Open Platform Communications Unified Platform. The Connect Edge gateway can be enabled as an OPC server for 10 or more devices.
network
You can connect 10 device or sensor adapters to each Connect Edge gateway. Gateways can host greater than 10 devices in many cases, however this is determined by reviewing post installation gateway CPU usage. Deciding which topology is applicable to your work or laboratory environment will determine the number of gateways you will need.
Connect Edge system has 3 main topologies
- Direct Wi-Fi-Device adapters are configured to transmit data directly to your Connect Edge gateway (10 per gateway). Your adapters must be in proximity (50-150 feet) to the gateway.
- Single/Dual Network- adapters are configured to connect to your local network via Ethernet or Wi-Fi (where permitted) and transmit data to the Connect Edge gateway, also on the same network. This topology is ideal if devices to be monitored are not within proximity to eachother or a gateway.
Building your system
- Determine type (Ethernet-only/WiFi+Ethernet) and quantity of gateway(s) needed based on network topology preference. The Configuration Planning Guide can be used for reference, if needed.
- Select compatible adapter for your lab equipment
- Purchase subscription/license for each device
- Determine whether self-install or professional-install is desired (where available)
System hardware and software is available for purchase in packages or a-la-carte. A built-in gateway discount is included with all packages. Additional savings with 3 and 5-year packages.
Installation
Customer (self) installation
Installation services sold separately, if desired (US, Europe, Great Britain, inquire with specialist for other regional availability).
Hardware Availability
Gateways
Connect Edge gateways are available in the following countries and configurations. For the most updated list, please ask your specialist.
Wi-Fi Connect Edge gateways are approved for these countries: US, CAN, UK, Europe, Australia, New Zealand, Colombia, Ecuador, Uruguay, Bolivia, Costa Rica, El Salvador, Guatemala, Puerto Rico, Thailand, Japan
Ethernet-only Connect Edge gateways are approved for these countries: Brazil, Honduras, Paraguay, Chile, Argentina, South Africa, Bahrain, Oman, Qatar, India, South Korea, Singapore, Vietnam
Adapters
Connect Edge adapter approved use varies by country. Please check the list to ensure adapters are approved for use in the desired country.
Wi-Fi Sensor Adapter/Device Adapter with power supply is approved for: US, CAN, Europe, UK, Australia, New Zealand, Colombia, Puerto Rico
Ethernet-only Sensor Adapter/Device Adapter with power supply is approved for: Ecuador, Uruguay, Bolivia, Costa Rica, El Salvador, Guatemala, Brazil, Honduras, Paraguay, Chile, South Africa, Bahrain, Oman, Qatar, Hong Kong
Adapter Kits exclude power supply for the countries below and can be purchased locally.
Wi-Fi Sensor Adapter/Device Adapter without power supply is approved for: India, Japan
Ethernet-only Sensor Adapter/Device Adapter without power supply is approved for: Thailand, Japan, South Korea, Singapore, Argentina, Vietnam
Local purchase specifications:
Device Adapter power supply specifications: Nominal: 5 VDC supply, 500mA (2.5W) max power consumption, peak power demand <2.5W
Sensor Adapter power supply specifications: Nominal: 5 VDC supply, 2A (10W) max consumption, peak power demand <15W
Specifications
| None | |
| InstrumentConnect | |
| 1 |