Learn More
Chemglass Inc 3mL Conical Reaction Vial, Minum-Ware, Graduated to 2mL
Supplier:  Chemglass Inc MW8004
Encompass
3mL Conical Reaction Vial, Minum-Ware, Graduated to 2mL. Heavy wall vial has an internal conical bottom for optimum sample retrieval and flat base which provides added mechanical strength and bench stability. Vials are graduated at the intervals listed. With 14/10 threaded outer joint. Supplied complete with open-top screw cap (MW-102-33), PTFE faced septa (MW-104-33) and Viton O-ring (MW-100-08).
Catalog No. MW8004
Only null left