Learn More
Alkali Scientific ASI™ Confocal Dish 35 x10mm, Clear, Easy-Grip, Insert Type, Sterile, PS/Glass, Non-Treated (Non-Charged/ Hydrophobic), 500/Cs
Catalog No. 503028616
Encompass
ASI™ product with a coverglass bottom is easy to use and facilitates preparation of cells for microscopic analysis. Optically clear with low background fluorescence coverglass was used. Ideal for use in high resolution and inverted microscopy, fluorescence imaging in live cells, confocal microscopy, phase contrast microscopy and micro-manipulations. Dish size : 35 mm Plate size: 6well plate(Cat.NO. 30106, 30206) Hole size : 13 or 20 Coverglass size : 22 x 22 mm(adhesion type, hole 13 ), 24 x 24(insert and adhesion type, hole 20 ) Coverglass thickness : 0.13 ~ 0.16 mm External grip 35 mm dish are available Black color 35mm dish is also available Sterilized by gamma irradiation Polystyrene
Catalog No. 503028616 Supplier Alkali Scientific Supplier No. TCG22
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ASI™ product with a coverglass bottom is easy to use and facilitates preparation of cells for microscopic analysis. Optically clear with low background fluorescence coverglass was used. Ideal for use in high resolution and inverted microscopy, fluorescence imaging in live cells, confocal microscopy, phase contrast microscopy and micro-manipulations. Dish size : 35 mm Plate size: 6well plate(Cat.NO. 30106, 30206) Hole size : 13 or 20 Coverglass size : 22 x 22 mm(adhesion type, hole 13 ), 24 x 24(insert and adhesion type, hole 20 ) Coverglass thickness : 0.13 ~ 0.16 mm External grip 35 mm dish are available Black color 35mm dish is also available Sterilized by gamma irradiation Polystyrene