missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Conductometer

Catalog No. S23665
Change view
Click to view available options
Grade:
7 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S23665 7 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S23665 Supplier Eisco™ Supplier No. PH0377

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Conductometer for demonstration of relative thermal conductivity of brass, copper, aluminum, iron, zinc and stainless-steel.

  • Rods of six metals radially spaced equally on brass hub.
  • Each rod with cavity at outer end for holding paraffin.
  • Brass hub held over flame for wax to melt at different times.

Specifications

Grade 7 to 12
For Use With (Application) demonstration of relative thermal conductivity of brass, copper, aluminum, iron, zinc and stainless-steel
Includes Rods of six metals radially spaced equally on brass hub
Product Type Conductometer