Learn More
Chemglass Inc Condenser, West, 365mm, 24/40 Joint, 200mm Jacket Length
Catalog No. CG122001
Encompass
Condenser, West, 365mm, 24/40 Joint, 200mm Jacket Length. Narrow annular space between jacket and inner tube of the West condenser provides for excellent cooling efficiency due to the increased flow rate of water. Condenser has a top standard taper outer joint and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration
Catalog No. CG122001 Supplier Chemglass Inc Supplier No. CG122001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Condenser, West, 365mm, 24/40 Joint, 200mm Jacket Length. Narrow annular space between jacket and inner tube of the West condenser provides for excellent cooling efficiency due to the increased flow rate of water. Condenser has a top standard taper outer joint and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration