Learn More
Chemglass Life Sciences Condenser, Reflux, 205mm, 14/20 Joint, Style A, 100mm Jacket Length DFS Item
Supplier:  Chemglass Inc CG121314
Condenser, Reflux, 205mm, 14/20 Joint, Style A, 100mm Jacket Length. Coil style reflux condenser with tightly wrapped coil providing maximum cooling. Excellent for use with high vapor pressure solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.
Catalog No. CG121314
Only null left
Explore available promotions