missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Condensate Drain High Performance Pipe Marker, White on Green

Catalog No. 18038096
Change view
Click to view available options
Length (English):
4 in.
10 in.
12 in.
16 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
18038096 4 in.
18038093 10 in.
18038095 12 in.
18038094 16 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18038096 Supplier Brady™ Supplier No. 5663OO

Strap-On High Performance Pipe Markers (B-689) are printed plastic sheets with sealed color graphics for use on pipes in harsh environments.

  • Designed for use on rusty, dirty, wet or rough pipes where pressure sensitive adhesive labels aren’t effective.
  • Resistant to UV light exposure, high humidity, high temperatures, abrasion, chemical reagents and physical agitation.
  • Excellent chemical resistance.
  • Durable corner grommets resist tears and rips to keep marker firmly in place using nylon ties.
  • Comes with two large nylon ties.

Specifications

Adhesive Type Non-adhesive
Coating Laminate
Color White on Green
Display Text
Finish Gloss
Format Sheet
Material Coiled Plastic (B-689)
Product Type Pipe Marker
Sterility Non-sterile
Dimensions (L x W) 4 x 4 in.
Length (English) 4 in.
Width (English) 4 in.
Width (Metric) 101.60 mm
Diameter (English) 0.375 to 0.5 in.
Show More Show Less