missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Concanavalin A
Description
Concanavalin A is a plant lectin that is a T-cell mitogen, and induces apoptosis in human fibroblasts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Concanavalin A is a plant lectin that is a T-cell mitogen, and induces apoptosis in human fibroblasts.
Solubility
Slightly soluble in water and phosfate buffered saline.
Notes
Incompatible with strong oxidizing agents.
Concanavalin A is a plant lectin that is a T-cell mitogen, and induces apoptosis in human fibroblasts.
Solubility
Slightly soluble in water and phosfate buffered saline.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Type | Concanavalin A |
| Physical Form | Lyophilized Powder |
| Color | White |
| Quantity | 250 mg |
| Merck Index | 14,2495 |
| Solubility Information | Slightly soluble in water and phosfate buffered saline. |
| Chemical Name or Material | Concanavalin A |
Safety and Handling
| Hazard Category | H317-H334 |
| Hazard Statement | GHS H Statement H334-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. |
| Precautionary Statement | P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c |
| EINECSNumber | 234-258-2 |
| RTECSNumber | GK6890000 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only