missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Concanavalin A (Con A) Conjugates

Catalog No. C860
Encompass
Change view
Click to view available options
Conjugate:
Alexa Fluor 350
Alexa Fluor 488
Alexa Fluor 594
Alexa Fluor 633
Alexa Fluor 647
Alexa Fluor Plus 405
Alexa Fluor Plus 750
Tetramethylrhodamine
Texas Red
Color:
Blue
Deep Red
Green
Near-infrared
Orange
Red
Violet
Excitation/Emission:
346/442 nm
405/450 nm
495/519 nm
555/580 nm
590/617 nm
595/615 nm
632/647 nm
650/668 nm
750/777 nm
Protein Subtype:
Concanavalin A (Con A)
Concanavalin A, succinylated
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate Color Excitation/Emission Protein Subtype
C860 Tetramethylrhodamine Orange 555/580 nm Concanavalin A (Con A)
C11254 Alexa Fluor 350 Blue 346/442 nm Concanavalin A (Con A)
C11252 Alexa Fluor 488 Green 495/519 nm Concanavalin A (Con A)
C21401 Alexa Fluor 488 Green 495/519 nm Concanavalin A, succinylated
C11253 Alexa Fluor 594 Red 590/617 nm Concanavalin A (Con A)
C21402 Alexa Fluor 633 Deep Red 632/647 nm Concanavalin A (Con A)
C21421 Alexa Fluor 647 Deep Red 650/668 nm Concanavalin A (Con A)
C56126 Alexa Fluor Plus 405 Violet 405/450 nm Concanavalin A (Con A)
C56127 Alexa Fluor Plus 750 Near-infrared 750/777 nm Concanavalin A (Con A)
C825 Texas Red Red 595/615 nm Concanavalin A (Con A)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. C860 Supplier Invitrogen™ Supplier No. C860

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates.

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates. Concanavalin A (Con A) has been used in a variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC) and flow cytometry (FC). In many cell types, concanavalin A (Con A) is a good marker of ER and Golgi.

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Flour and Alexa Flour Plus conjugates. Concanavalin A selectively binds to alpha‐mannopyranosyl and alpha‐glucopyranosyl residues. In neutral and alkaline solutions, concanavalin A exists as a tetramer with a molecular weight of approximately 104,000 Daltons. In acidic solutions (pH below 5.0), concanavalin A exists as a dimer. Conjugated concanavalin A has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).

Specifications

For Use With (Application) Immunocytochemistry, Immunohistochemistry, Immunofluorescence
Quantity 10 mg
Source Canavlia ensiformis
Conjugate Tetramethylrhodamine
Cross Reactivity α-man, α-glc
Species C. ensiformis
Recombinant Native
Protein Tag None
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Excitation/Emission 555/580 nm
Label Type Classic Dyes
Shipping Condition Room Temperature
Expression System Natural
Protein Family Lectins
Form Lyophilized
Protein Subtype Concanavalin A (Con A)
Research Category Imaging
Color Orange
Purity or Quality Grade Imaging Quality
Ligand Type Lectin
Show More Show Less