Learn More
Invitrogen™ Concanavalin A (Con A) Conjugates
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates.
Supplier: Invitrogen™ C56127
Description
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates. Concanavalin A (Con A) has been used in a variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC) and flow cytometry (FC). In many cell types, concanavalin A (Con A) is a good marker of ER and Golgi.
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Flour and Alexa Flour Plus conjugates. Concanavalin A selectively binds to alpha‐mannopyranosyl and alpha‐glucopyranosyl residues. In neutral and alkaline solutions, concanavalin A exists as a tetramer with a molecular weight of approximately 104,000 Daltons. In acidic solutions (pH below 5.0), concanavalin A exists as a dimer. Conjugated concanavalin A has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).Chemical Identifiers
| Immunocytochemistry, Immunohistochemistry, Immunofluorescence | |
| α-man, α-glc | |
| Native | |
| Room Temperature | |
| Lyophilized | |
| Imaging Quality |
| Canavlia ensiformis | |
| C. ensiformis | |
| None | |
| Lectins | |
| Imaging | |
| Lectin |
Specifications
| Immunocytochemistry, Immunohistochemistry, Immunofluorescence | |
| Canavlia ensiformis | |
| α-man, α-glc | |
| Native | |
| Store in freezer (-5°C to -30°C) and protect from light. | |
| Classic Dyes | |
| Lectins | |
| Concanavalin A (Con A) | |
| Near-infrared | |
| Imaging Quality |
| 1 mg x 5 vials | |
| Alexa Fluor Plus 750 | |
| C. ensiformis | |
| None | |
| 750/777 nm | |
| Room Temperature | |
| Lyophilized | |
| Imaging | |
| Alexa Fluor | |
| Lectin |