Learn More
Invitrogen™ Concanavalin A (Con A) Conjugates
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates.
Supplier:  Invitrogen™ C21421
Encompass
 View more versions of this product
Catalog No. C21421
Only null left
Description
Description
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates. Concanavalin A (Con A) has been used in a variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC) and flow cytometry (FC). In many cell types, concanavalin A (Con A) is a good marker of ER and Golgi.
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Flour and Alexa Flour Plus conjugates. Concanavalin A selectively binds to alpha‐mannopyranosyl and alpha‐glucopyranosyl residues. In neutral and alkaline solutions, concanavalin A exists as a tetramer with a molecular weight of approximately 104,000 Daltons. In acidic solutions (pH below 5.0), concanavalin A exists as a dimer. Conjugated concanavalin A has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).
Specifications
Chemical Identifiers
Immunocytochemistry, Immunohistochemistry, Immunofluorescence
α-man, α-glc
Native
Room Temperature
Lyophilized
Imaging Quality
Canavlia ensiformis
C. ensiformis
None
Lectins
Imaging
Lectin
Specifications
Immunocytochemistry, Immunohistochemistry, Immunofluorescence
Canavlia ensiformis
α-man, α-glc
Native
Store in freezer (-5 to -30°C) and protect from light.
Alexa Fluor Dyes
Natural
Lyophilized
Imaging
Alexa Fluor
Lectin
5 mg
Alexa Fluor 647
C. ensiformis
None
650/668 nm
Room Temperature
Lectins
Concanavalin A (Con A)
Deep Red
Imaging Quality
Product Suggestions
Product Suggestions