missing translation for 'onlineSavingsMsg'
Learn More

COMU™, 97%

Catalog No. p-4258911
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC439480050 5 g Glass bottle
AC439480100 10 g Glass bottle
AC439481000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC439480050 Supplier Thermo Scientific Chemicals Supplier No. 439480050
Only null left

Coupling reagent | CAS: 1075198-30-9 | C12H19F6N4O4P | 428.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 1075198-30-9
Molecular Formula C12H19F6N4O4P
Molecular Weight (g/mol) 428.27
MDL Number MFCD11975052 MFCD24368564 MFCD11975052
InChI Key GPDHNZNLPKYHCN-UHFFFAOYSA-N
Synonym comu, 1-cyano-2-ethoxy-2-oxoethylidenaminooxy dim, 1-cyano-2-ethoxy-2-oxoethylidenaminooxy dimethylamino-morpholinocarbenium hexafluorophosphate, 1-cyano-2-ethoxy-2-oxoethylideneaminooxy-dimethylamino-morpholino-carbenium hexafluorophosphate, z-1-cyano-2-ethoxy-2-oxoethylidene amino oxy-n,n-dimethyl morpholin-4-yl methaniminium hexafluorophosphate, 4-e-1-cyano-2-ethoxy-2-oxoethylidene amino oxidanyl dimethylamino methylidene-4??-morpholin-4-ylium hexafluorophosphate
PubChem CID 44471148
IUPAC Name [[(Z)-(1-cyano-2-ethoxy-2-oxoethylidene)amino]oxy-morpholin-4-ylmethylidene]-dimethylazanium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.CCOC(=O)C(=NOC(N1CCOCC1)=[N+](C)C)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 151.4°C to 152.9°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 428.27
Percent Purity 97%
Physical Form Powder or Solid
Chemical Name or Material COMU*
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only