Learn More
Merck Emd Millipore cOmplete, Mini Protease Inhibitor Cocktail, 25 tablets
Supplier:  Merck Emd Millipore 11836153001
Encompass
Has been used as a component of: Lysis buffer for lysing glioma tumor samples, radioimmunoprecipitation (RIPA) protein extraction buffer for homogenization of heart tissues, resuspension buffer for retinal ganglion cells homogenization. Tablets provided in a glass vial.
Catalog No. NC1377710
Only null left