Learn More
Merck Emd Millipore cOmplete™, Mini, EDTA-free Protease Inhibitor Cocktail
Supplier:  Merck Emd Millipore 11836170001
Encompass
Protease Inhibitor Cocktail Tablets provided in a glass vial ; Easy-to-use: Simply drop a quick-dissolving tablet into your buffer. ; Safe: Choose non-toxic inhibitors that pose no risk to you or those around you. ; Complete protection: Instantly protect your proteins against a broad range of proteases. ; Flexible: Protect proteins in extracts from almost any tissue or cell, including animals, plants, yeast, bacteria, or fungi.
Catalog No. NC0962311