missing translation for 'onlineSavingsMsg'
Learn More

CEP Complete Environmental Products™ 65 Gallon Poly Overpack Salvage Drum

Catalog No. p-4706961
Change view
Click to view available options
Diameter (English) Outer:
28.25 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Outer
19087802 28.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19087802 Supplier CEP Supplier No. 1065 YE
Only null left

Specifications

Certifications/Compliance Meets Group I packaging standards
Closure Type Gasket Lid
Color Yellow
For Use With (Equipment) Fits 30 gal. Drums
Diameter (English) Outer 28.25 in.
Weight (English) 47 lbs.
Weight (Metric) 21 Kg