missing translation for 'onlineSavingsMsg'
Learn More

Levocarnitine, Free Base, USP, 97-103%, Spectrum™ Chemical

Catalog No. 18603806
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603806 25 kg Fiber Drum
18603801 25 g Poly Bottle
18603802 100 g Poly Bottle
18603803 500 g Poly Bottle
18603804 1 kg Poly Pail
18603805 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18603806 Supplier Spectrum Chemical Mfg Cor Supplier No. C139925KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

C1399, 541-15-1, C7H15NO3

Spectrum™ Chemical Levocarnitine, Free Base, USP is used to prevent and treat a lack of carnitine. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 541-15-1
Molecular Formula C7H15NO3
Molecular Weight (g/mol) 161.20
InChI Key PHIQHXFUZVPYII-UHFFFAOYNA-N
IUPAC Name 3-hydroxy-4-(trimethylazaniumyl)butanoate
SMILES C[N+](C)(C)CC(O)CC([O-])=O

Specifications

CAS Max % 100%
Quantity 25 kg
Ignition Residue 0.5%
Packaging Fiber Drum
Percent Purity 97 to 103%
Grade USP