New England Biolab Ltd SHuffle T7 Competent E.coli 12 x 0.05 ml/tube
Catalog No. NC1323515
Chemically competent E. coli K12 cells suitable for T7 protein expression with enhanced capacity to correctly fold proteins with multiple disulfide bonds in the cytoplasm.
Catalog No.
NC1323515
Supplier
New England Biolab LtdSupplier No.
C3026J
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Chemically competent E. coli K12 cells suitable for T7 protein expression with enhanced capacity to correctly fold proteins with multiple disulfide bonds in the cytoplasm.