missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Compact Ear Plug Dispenser

Catalog No. p-7214047
Change view
Click to view available options
Product Type:
With cover
Without cover
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19104415 With cover
19104414 Without cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19104415 Supplier Brady™ Supplier No. 2019L
Only null left

Acrylic compact dispensers serve as a visual reminder for employees to wear their ear plugs. Conveniently mount near hazardous work areas to encourage compliance.

  • Clear acrylic with green HEARING PROTECTION text legend
  • Can be wall or table mounted for convenient access
  • Size is 3 X 9 in. with a 6 in. back plate
  • Available with or without top cover
  • Ear plugs not included

Specifications

Depth (English) 6 in.
Depth (Metric) 15.2 cm
For Use With (Equipment) Ear plugs
For Use With (Application) Ear Plug Dispenser with Visual Reminder for Employees to Wear Ear Plugs When Mounted Near Hazardous Work Areas.
Height (English) 3 in.
Height (Metric) 7.6 cm
Holds Ear plugs
Width (Metric) 22.8 cm
Width (English) 9 in.
Product Type With cover
Disposable Reusable
Material Acrylic
Mounting Wall Mountable
Show More Show Less