missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Compact Bucket System

Catalog No. 19169117
Change view
Click to view available options
Material:
Polypropylene
Product Type:
Standard Handle
Utility Handle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type
19169117 Polypropylene Utility Handle
19169116 Polypropylene Standard Handle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19169117 Supplier Contec™ Supplier No. 2940U2B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contec Compact Bucket System is an all-in-one solution to make cleaning and disinfecting easier and more efficient

  • Designed for use with Contec mops such as the VertiKlean™ MAX™, VertiKlean™ MAX™ Sealed Edge, EasyCurve™ and QuickTask™ Mopping Systems
  • Stainless steel cart is available in multiple configurations to meet the unique needs of the environment
  • Available with either two or three, autoclavable polypropylene buckets, or with three stainless steel buckets, and a choice of a standard or a utility cart handle
  • Utility handle allows two small buckets (6 L) or one large bucket (15 L) to be positioned on the handle providing additional storage for extra supplies such as clean mop heads, solution spray bottles, wipes, and other items as needed
  • Cart frame has optional mop head removal tool (model no. 2639) on the front for easy, hands-free removal of a mop head
  • Two mop holders with handle clasps on either side of the cart allow for convenient mop storage during transport and storage
  • Polypropylene buckets come with a stainless steel debris screen positioned in the base of the clean and rinse buckets with a three bucket system and in the clean bucket for a two bucket system, preventing the mop from touching the bottom of the bucket where unwanted contaminants settle

Specifications

Product Type Utility Handle
Material Polypropylene
Autoclavable Autoclavable
Capacity (English) 3 x 3.9 gal.
Color Red, Black and Blue
Disposable Reusable
For Use With (Application) Cleaning and disinfecting
For Use With (Equipment) Contec mops such as the VertiKlean™ MAX™, VertiKlean™ MAX™ Sealed Edge, EasyCurve™ and QuickTask™ Mopping Systems
Quantity 1 Ea.