missing translation for 'onlineSavingsMsg'
Learn More

3M™ Comfort Belt, CB-1000

Catalog No. 296367842
Change view
Click to view available options
1/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
296367842 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 296367842 Supplier 3M™ Supplier No. 7000126406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Adjustable belt that is designed to evenly distribute the weight of a 3M™ Belt-Mounted Powered Air Purifying Respirator GVP-Series. The 3M™ Comfort Belt, CB-1000 can accommodate waist sizes from 26 in x 54 in (66.04 cm x 137.16 cm).

  • Delve into 3M™ Products and experience the wide variety of products and replacement parts offered with powered air purifying respirators (PAPR)
  • In addition to powered systems, 3M also offers supplied air respirators (SAR) using air from an air tank or a different external source

Specifications

Color Black
Material Leather
Product Type Belt
For Use With (Application) Used in applications such as assembly, blasting, cleaning, metal repair, painting
Quantity 1 Ea.