missing translation for 'onlineSavingsMsg'
Learn More

Dentec Safety™ Comfort-Air™ Series 300 Half Mask Respirators

Catalog No. 19222414
Change view
Click to view available options
Size:
Medium/Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19222414 Medium/Large
19222413 Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19222414 Supplier Dentec Safety™ Supplier No. DTC6516435ML00

Thermoplastic half-mask respirators provide comfort and durability with a flexible sealing edge and multi-position cartridge attachment.

  • Extended field of vision
  • Large inhalation valves
  • Exhalation valve is strategically positioned under the nostrils and in front of the mouth to ensure hot, exhaled air exits the mask immediately
  • Textured sealing area reduces risk of the mask slipping on the face to ensure a comfortable seal on a wide variety of face sizes
  • Enhanced harness assembly features:
    • Metal tabs at end to prevent nylon strap fraying and allowing for adjustment when wearing gloves
    • Wide nylon strap is more comfortable against the skin and maintains its elasticity longer
    • 4-point swiveling adjustments allows the mask to be adjusted
    • Cradle suspension fits comfortably under suspension systems for hard hats, grinding and welding helmets
  • 3-position adjustment allows users to adjust cartridge to three different positions

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Gray
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Medium/Large
With Exhalation Valve Yes
Facepiece Material Thermoplastic Elastomer
For Use With (Equipment) Dentec™ Filters and Cartridges
Model 300-ML-00
Length (Metric) 8.89 cm
Width (Metric) 17.78 cm
Height (Metric) 12.7 cm
Show More Show Less