missing translation for 'onlineSavingsMsg'
Learn More

Dentec Safety™ Comfort-Air™ Series 100 Half-Mask Respirators

Catalog No. 19222410
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19222410 Small
19222411 Medium
19222412 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19222410 Supplier Dentec Safety™ Supplier No. DTC6516417S00

Pure silicone half-mask respirators are comfortable and easy to clean.

  • Extended field of vision
  • Large inhalation valves
  • Exhalation valve is strategically positioned under the nostrils and in front of the mouth to ensure hot, exhaled air exits the mask immediately
  • Extra Large face seal surface offers comfort
  • Textured finish on the sealing area reduces the risk of the mask slipping on the face
  • Enhanced harness assembly features:
    • Metal tabs at the end to prevent nylon strap from fraying and allowing for adjustment when wearing gloves
    • Wide nylon strap maintains elasticity longer.
    • 4-point swiveling adjustments
    • Cradle suspension fits comfortably under suspension systems for hard hats, grinding and welding helmets
  • 3-position adjustment allows users to adjust cartridge to three different positions

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Blue
Material Silicone
Product Type Half-Mask Respirator
Size Small
With Exhalation Valve Yes
Facepiece Material Silicone
For Use With (Equipment) Dentec™ Filters and Cartridges
Material Headstrap Nylon
Model 100-S-00
Length (Metric) 8.89 cm
Width (Metric) 17.78 cm
Height (Metric) 12.7 cm
Show More Show Less