missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ CometAssay Alkaline Control Cells
Description
- Stored using a Liquid Nitrogen Storage System
- Includes:
- 500 μL of Healthy Control Cells
- 500 μL of Low Damage Control Cells
- 500 μL of Intermediate Damage Control Cells
- 500 μL of High Damage Control Cells
- These cryopreserved controls are designed to act as controls to standardize and compare alkaline electrophoresis methods between individual users and laboratories
Specifications
Specifications
| Quantity | 10 Tests |
| Synonym | Apoptosis kinetics, apoptosis kit, apoptosis kit annexin v, apoptosis kit microscopy |
| Content And Storage | Store in liquid nitrogen. Do not use past expiration date. |
| Product Line | R&D Systems |
| For Use With (Application) | Controls with CometAssay™ Kit |