missing translation for 'onlineSavingsMsg'
Learn More

Combretastatin A4 Phosphate Disodium Salt 98.0+%, TCI America™

Catalog No. p-7125778
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
20 mg
Catalog No. Quantity
C300920MG 20 mg
C3009100MG 100 mg
2 options

Catalog No. C300920MG

Supplier: TCI America C300920MG

Only null left

Chemical Identifiers

CAS 168555-66-6
Molecular Formula C18H25Na2O8P
Molecular Weight (g/mol) 446.343
MDL Number MFCD00943911
InChI Key UGAFMFUUPVSHLZ-YGGCHVFLSA-N
Synonym Fosbretabulin Disodium Salt
PubChem CID 131673796
IUPAC Name [2-methoxy-5-[(Z)-2-(3,4,5-trimethoxyphenyl)ethenyl]phenyl] dihydrogen phosphate;molecular hydrogen;sodium
SMILES [HH].[HH].COC1=C(C=C(C=C1)C=CC2=CC(=C(C(=C2)OC)OC)OC)OP(=O)(O)O.[Na].[Na]

Specifications

Color White
Quantity 20 mg
Formula Weight 440.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Combretastatin A4 Phosphate Disodium Salt
TSCA No
Recommended Storage Freezer