missing translation for 'onlineSavingsMsg'
Learn More

Combretastatin A4 Phosphate Disodium Salt 98.0+%, TCI America™

Catalog No. C300920MG
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C300920MG 20 mg
C3009100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C300920MG Supplier TCI America Supplier No. C300920MG

Chemical Identifiers

CAS 168555-66-6
Molecular Formula C18H25Na2O8P
Molecular Weight (g/mol) 446.343
MDL Number MFCD00943911
InChI Key UGAFMFUUPVSHLZ-YGGCHVFLSA-N
Synonym Fosbretabulin Disodium Salt
PubChem CID 131673796
IUPAC Name [2-methoxy-5-[(Z)-2-(3,4,5-trimethoxyphenyl)ethenyl]phenyl] dihydrogen phosphate;molecular hydrogen;sodium
SMILES [HH].[HH].COC1=C(C=C(C=C1)C=CC2=CC(=C(C(=C2)OC)OC)OC)OP(=O)(O)O.[Na].[Na]

Specifications

Color White
Quantity 20 mg
Formula Weight 440.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Combretastatin A4 Phosphate Disodium Salt
TSCA No
Recommended Storage Freezer