missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Combitips™ Advanced Empty Box 2.0

Catalog No. 14380058
Change view
Click to view available options
Volume (Metric):
0.1 to 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14380058 0.1 to 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14380058 Supplier Eppendorf™ Supplier No. 0030089952

Streamline your workflow with the Eppendorf Combitips™ advanced Empty Box 2.0. This reusable box stores 8 Combitips™ advanced, ensuring easy access and quick tip attachment, keeping your lab organized and efficient.

  • Reusable box to safely store 8 Combitips™ advanced (0.1 – 10 mL) for easy tip attachment.
  • Autoclavable at 121°C (20 min.) for subsequent use.

Specifications

Description Empty box
Volume (Metric) 0.1 to 10 mL
Color Gray
Autoclavable Autoclavable
Format Box
Tip Style Positive Displacement
Product Type Empty Box
Disposable Reusable
For Use With (Equipment) Eppendorf Combitips
Product Line CombiTips Advanced