Learn More
BUCHI Combi-Clip
For use with Rotavapor R-300
Supplier:  BUCHI 11059770
Catalog No. 05405053
Only null left
Explore available promotions
Description
Description
  • Patented Combi-Clip with snap lock mechanism
  • For fastening evaporating flask on to vapor duct
  • Vapor duct sold separately
Specifications
Specifications
PBT
Combi Clip
Fastening Evaporating Flask on Vapor Duct
Rotavapor R-300
Product Suggestions
Product Suggestions