missing translation for 'onlineSavingsMsg'
Learn More

Comanic Acid 98.0+%, TCI America™

Catalog No. C11041G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11041G 1 g
C11045G 5 g
C110425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C11041G Supplier TCI America Supplier No. C11041G
Only null left

Chemical Identifiers

CAS 499-05-8
Molecular Formula C6H4O4
Molecular Weight (g/mol) 140.094
MDL Number MFCD00059958
InChI Key UFMDRPPBUAPWBK-UHFFFAOYSA-N
Synonym 4-Oxo-4H-pyran-2-carboxylic Acid, gamma-Pyrone-2-carboxylic Acid
PubChem CID 12305536
IUPAC Name 4-oxopyran-2-carboxylic acid
SMILES C1=COC(=CC1=O)C(=O)O

Specifications

Quantity 1 g
Formula Weight 140.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Comanic Acid