Learn More
Microsolv Technology Column Diamond Hydr 2.1X150Mm DFS Item
Supplier:  Microsolv Technology Corporation 7000015P2
HPLC Column, Diamond Hydride, 4um, 2.1mm ID x 150mm Length, 100A. Cogent TYPE-C Brand. 1/EA.
Catalog No. 50-014-51322
Only null left
Explore available promotions