missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Color Filter Sets
Description
Variety of colors individually mounted in 2 x 2 in. (5 x 5cm) 35mm plastic slide holders.
Basic Color Filter Set:
- Three filters: red, blue, green
Advanced Color Filter Set:
- 14 filters: light blue, medium blue, daylight blue, dark blue, golden amber, medium green, light green, green-blue, light red, medium purple, orange, medium yellow, medium red, frost
Specifications
Specifications
| Product Type | Color Filter Set |
| For Use With (Application) | Ideal for use in optical transmission measurements, solar monitoring and exploration, attenuation of laser beams and demonstrations of laser light's monochromacity |
| Includes | Three filters: one red, blue and green |