missing translation for 'onlineSavingsMsg'
Learn More

Colloidal Silicon Dioxide, NF, EP, 99-100.5%, Spectrum™ Chemical

S1510, 7631-86-9, SiO2

Supplier:  Spectrum Chemical Mfg Cor S15105KGBL

 View more versions of this product

Catalog No. 18601821


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Colloidal Silicon Dioxide, NF, EP is used both in the food and pharmaceutical industries as a powder flow helper in the production of tablets. All Spectrum Chemical NF, EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

Colloidal Silicon Dioxide
100%
5 kg
2.5%
5%
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
99 to 100.5%
7631-86-9
Silicon Dioxide
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Fiber Drum
O=[Si]=O
60.08
Multi-Compendial

Chemical Identifiers

7631-86-9
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
O=[Si]=O
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
silanedione
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions