missing translation for 'onlineSavingsMsg'
Learn More

Collodion, USP, Spectrum™ Chemical

Catalog No. 18601815
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
200 L
Packaging:
Amber Glass Bottle
Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601815 500 mL Amber Glass Bottle
18601814 100 mL Amber Glass Bottle
18601816 4 L Amber Glass Bottle
18601817 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601815 Supplier Spectrum Chemical Mfg Cor Supplier No. CO120500MLGL
Only null left

CO120, 9004-70-0

Spectrum™ Chemical Collodion, USP is used as a surgical dressing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-70-0
Molecular Formula C24H36N8O38
Molecular Weight (g/mol) 1044.57
InChI Key OWWGYSXLINVAES-UHFFFAOYNA-N
IUPAC Name 2,3-bis(nitrooxy)-6-[(nitrooxy)methyl]-5-{[3,4,5-tris(nitrooxy)-6-[(nitrooxy)methyl]oxan-2-yl]oxy}oxan-4-yl nitrate; 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O.[O-][N+](=O)OCC1OC(O[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1OC1OC(CO[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1O[N+]([O-])=O

Specifications

CAS Max % 100%
UN Number UN2059
Quantity 500 mL
Percent Purity 5%
Grade USP
Packaging Amber Glass Bottle