missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Collisions Kit

Catalog No. S15118
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S15118 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15118 Supplier Eisco™ Supplier No. PH0352CKT

The Collisions Kit is an ideal tool to supplement introducing students to the concept of collision, momentum, and conservation of energy.

  • Ideal for studying collision in terms of momentum and conservation of energy
  • Instruction booklet is complete with a Teacher's Guide and reproducible Student Guide
  • Recommended for Grades 9-12

Kit Includes

  • Collision Track, 2 Collision Carts, 6 Slotted Masses, 2 Support Feet with Leveling Screws, 2 Photogate Support Rods, 2 Support Rod Holders, 1 Pulley, and Instruction Booklet

Required (Not Included)

  • Photogate System, Mass Hanger, Slotted Weights, Balance, Masking Tape, String, Level, Meter Stick, Stopwatch, Protractor, Books, and Graph Paper

Specifications

Product Type Collisions Kit
Grade 7 to 14
Size 1.2 m (Track)
For Use With (Application) Supplementing the Concepts of Collision, Momentum, and Energy Conservation
Includes Collision Track, 2 Collision Carts, 2 Sets of 3 Slotted Masses (10 g, 20 g, and 50 g), 2 Support Feet with Leveling Screws, 2 Photogate Support Rods, 2 Support Rod Holders, 1 Pulley, and Instructions