Learn More
Waters Limited 96-well Sample Collection Plate, 350 L, 50/pk
Catalog No. 50818657
Encompass
96-well sample collection plate with 350 L well. This plate is SBS/ANSI compliant for robot compatible systems. This plate can also serve as a collection plate for 96-well SPE and filtration-plate formats.
Catalog No. 50818657 Supplier Waters Limited Supplier No. WAT058943
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
96-well sample collection plate with 350 L well. This plate is SBS/ANSI compliant for robot compatible systems. This plate can also serve as a collection plate for 96-well SPE and filtration-plate formats.