Learn More
Eisco™ Collapsable Metal Can
For demonstration of ideal gas law physics principles relating to gas and air pressure.
Supplier: Eisco™ PH0201602
Description
- With stopper and hose fitting
- For use with vacuum hand pump
- Can be collapsed using boiling water to evacuate air from can
Specifications
| Collapsable Can | |
| Metal | |
| Demonstration of ideal gas law physics principles relating to gas and air pressure | |
| Cap, stopper, and hose fitting |