Learn More
Chrono Log Corp CHRONO-PAR Collagen Reagent, High Concentration Platelet Aggregation Reagent, Validated for Whole Blood and PRP, 1 mL
Supplier: Chrono Log Corp 385
The CHRONO-PAR Collagen Reagent is a highly concentrated platelet aggregation reagent that enables researchers to conduct detailed platelet function and secretion studies. It efficiently activates platelets with 1 mg of native Type I collagen sourced from equine tendons. This versatile reagent is validated for use with various blood samples, making it essential for diagnosing platelet dysfunctions and confirming normal platelet function in diagnostic applications.