missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Collagen I, rat tail

Numéro de catalogue. A1048301
Encompass
Modifier l'affichage
Click to view available options
Quantité:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
A1048301 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. A1048301 Fournisseur Gibco™ Code fournisseur A1048301

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Collagen I Rat Protein, Tail is the most common fibrillar collagen (90%) found in skin, bone, tendons, and other connective tissues.

Collagen I, Rat Tail is the most common fibrillar collagen (90%) found in skin, bone, tendons, and other connective tissues. It is supplied at ~4 mg/mL in 20 mM acetic acid, providing the flexibility to dilute to lower concentrations. (See Certificate of Analysis for exact concentration.) The detailed gelling procedure forms a firm, clear gel for 3D applications.

Collagen is a fibrous protein that consists of three α-chains that can combine to form a rope-like triple helix, providing tensile strength to the extracellular matrix (ECM) where it plays a key role in cell growth, differentiation, attachment, and cell migration. The α-chains contain GXY repeats. Glycine (G) is a small amino acid that fits well in the triple helix. X and Y are typically proline and hydroxyproline, which is critical for collagen stability. Type I is the most common fibrillar collagen (90%) and is mostly found in skin, bone, tendons, and other connective tissues.

Rat tail collagen I can be prepared as a clear gel providing a 3D matrix or surface coated on tissue culture plates as a substrate for culturing primary cells such as keratinocytes and hepatocytes.

Spécifications

Concentration 4 mg/mL
À utiliser avec (application) Cell Culture
Quantité 25 mL
Emballage Plastic Bottle
Type de produit Collagen
Conjugué Unconjugated
Espèces Rat
Recombinant Native
Stérilité Sterile
Contenu et stockage Store in refrigerator (2–8°C) and protect from light.
Taux d’endotoxine ≤10 EU/mL, ≤ 10 EU/mL
Shipping condition (Condition d'expédition) Wet Ice
Forme de protéine Native
Forme Liquid
Sous-type de protéine Collagen
Catégorie de recherche Extracellular Matrix & Cell Adhesion
Couleur Clear
Gamme de produit Gibco
Afficher plus de résultats Afficher moins de résultats

For Research Use Only. Not for use in diagnostic procedures.